Learn More
Medchemexpress LLC Fluo-3-pentaacetoxymethyl ester | 121714-22-5 | ≥99.0% | 1 MG
SDP
Supplier:  Medchemexpress LLC HYD07161MG
Encompass_Preferred
Fluo-3 AM is a fluorescent Ca2+ chelator with high affinity for calcium. It can specifically identify intracellular calcium ions, exhibiting high sensitivity and low cytotoxicity. The increased AM acetylmethyl ester allows it to enter the cell effectively, and after being cleaved by intracellular esterase, it remains in the cell to bind with calcium ions, producing strong fluorescence.
  • Fluorescent Ca2+ chelator.
  • High affinity for calcium.
  • Specifically identifies intracellular calcium ions.
  • High sensitivity.
  • Low cytotoxicity.
  • Increased AM acetylmethyl ester for good cellular penetration.
  • Produces strong fluorescence upon binding to calcium ions.
  • Can be used in calcium flux analysis.
Catalog No. 50-004-10482