Learn More
Biotium Fluo-3, Pentaammonium Salt
Supplier:  Biotium 50010
Encompass_Preferred
Fluo-3, pentaammonium is membrane-impermeant, but can be loaded into cells via microinjection or scrape loading. Kd: 450 nM Properties: Ex/Em (low or high [Ca2 ]) = 506nm/526nm; e; (506 nm) = 86,000 M-1cm-1; Orange red solid soluble in DMSO and water (pH >;6); Store at 4°C and protect from light, especially when in solution; C36H25Cl2N7O13; MW: 854.7; [134907-84-9]
Catalog No. 50-196-4684