Learn More
Medchemexpress LLC Fluo-4 AM | 273221-67-3 | MFCD18837018 | 97.7% | 1096.94 g·mol⁻¹ | C51H50F2N2O23 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1018965MG
Encompass_Preferred
Fluo-4 AM is a cell-permeable acetoxymethyl (AM) ester calcium indicator that is hydrolyzed by intracellular esterases to release fluorescent Fluo-4. It enables measurement of intracellular Ca2+ dynamics in live cells, and is suitable for fluorescence microscopy, plate readers, flow cytometry, and high-throughput screening (Ex 485 nm / Em 526 nm).
  • Cell-permeable AM ester for live-cell calcium imaging.
  • Bright green fluorescence upon Ca2+ binding (Ex 485 nm, Em 526 nm).
  • Compatible with microscopy, plate readers, and flow cytometry.
  • Cleaved by intracellular esterases to yield active fluorescent dye.
  • Available in multiple package sizes for varied experimental scales.
Catalog No. 50-002-48912