Learn More
Apexbio Technology LLC Fluo-4 AM Calcium Assay Kit 1000tests
SDP
Supplier:  Apexbio Technology LLC K22981000
Encompass_Preferred
Fluo-4 AM is a cell-permeant fluorescent calcium indicator widely used for detecting intracellular calcium dynamics Upon cellular entry endogenous esterases cleave the acetoxymethyl (AM) ester groups yielding the active Fluo-4 dye which exhibits strong fluorescence enhancement upon binding free Ca ions This property enables sensitive real-time monitoring of calcium signaling in live cells by fluorescence microscopy flow cytometry or microplate assays The Fluo-4 AM Calcium Assay Kit is suitable for applications in pharmacological screening cellular physiology studies and analyses of receptor-mediated calcium flux in a variety of cell types
Catalog No. 50-000-96993