Learn More
Medchemexpress LLC Fluoflavine | 531-46-4 | 99.4% | 234.26 g/mol | C14H10N4 | 25MG

Supplier: Medchemexpress LLC HY12619525MG
Fluoflavine is a small-molecule research compound that selectively inhibits NADPH oxidase 1 (NOX1). It is used in biochemical and cellular studies to probe NOX1-mediated reactive oxygen species production. The compound exhibits potent enzymatic inhibition and demonstrable cellular activity, and is supplied in small milligram quantities with high analytical purity.
- Selective NOX1 inhibition with IC50 of 90 nM.
- Greater than 100-fold selectivity over NOX2, NOX3, and NOX4 (IC50 >10 μM).
- Cellular activity: IC50 of 360 nM in HEK293 cells.
- High analytical purity (99.41%).
- Available in small milligram pack sizes suitable for research assays.
- Molecular formula C14H10N4 and molecular weight 234.26 g/mol.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.