Learn More
Enzo Life Sciences FLUOR DE LYS-Succinyl (50µl)
SDP
Supplier:  Enzo Life Sciences BMLKI5900050
Encompass_Preferred
The FLUOR DE LYS-Succinyl, Desuccinylase Substrate offers a highly sensitive and convenient alternative to radiolabeled, succinylated histones or peptide/HPLC methods for the assay of desuccinylases such as SIRT5.  The assay procedure has two steps.  First, the FLUOR DE LYS- Succinyl, Desuccinylase Substrate, which comprises a succinylated lysine side chain, is incubated with a sample containing desuccinylase activity (HeLa nuclear or other extract, purified enzyme, bead-bound immunocomplex, etc.).  Desuccinylation of the substrate sensitizes the substrate so that, in the second step, treatment with the FLUOR DE LYS Developer (BML-KI105) produces a free AMC fluorophore which can be monitored with standard fluorometers (Ex. 350-380nm / Em. 450-480nm). Formulation: Solution in DMSO. Purity: ≥95% (HPLC). Long Term Storage: -80°C.
Catalog No. 50-201-1126