Learn More
Cayman Chemical FluorescamIn 500mg
SDP
Supplier:  Cayman Chemical 34286500 MG
Encompass_Preferred
An amine-reactive fluorescent probe; has commonly been used as a reagent in fluorometric protein assays; has commonly been used as a reagent in fluorometric protein assays; displays excitation/emission maxima of 370/488 nm, respectively, in acetone
Catalog No. 50-294-8937