Learn More
TARGETMOL CHEMICALS INC Fluorescein-5-maleimide 25MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T1899125MG
Encompass_Preferred
Also available in 1 mL, 5 mg, 10 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Fluorescein-5-maleimide (N-(5-Fluoresceinyl)maleimide) is a fluorescent thiol-reactive dye (excitation 494 nm, emission 519 nm). Purity 97.42%
Catalog No. 50-287-6902