Learn More
Cayman Chemical FluoresceIn-5-maleImIde 25mg
SDP
Supplier:  Cayman Chemical 1638325 MG
Encompass_Preferred
An activated fluorescent molecule (excitation: 494 nm, emission: 519 nm) used for labeling proteins; reacts optimally with sulfhydryl groups on cysteine side chains at pH 7, forming a stable thioether bond
Catalog No. 50-297-1016