Learn More
Abcam Fluorescein-5-thiosemicarbazide, Fluorescent tag, 100MG
Supplier:  Abcam AB145390100MG
Encompass_Preferred
MW 421.43 g/mol, Purity >97%. Fluorescent tag for labeling of cell-surface functional groups (glycophorins). Also used as a probe for detecting protein carbonyl derivatives.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5235