Learn More
Abcam Fluorescein di-ß-D-glucuronide, dimethyl ester, 2MG
Supplier:  Abcam AB2755442MG
Encompass_Preferred
MW 712.61 g/mol, Purity >95%. Sensitive, cell-permeant β-glucuronidase substrate for live cell and tissue assays of cloned GUS marker gene activity and transfection levels.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-268-2774