Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Fluorescein di-β-D-galactopyranoside | 17817-20-8 | MFCD00077344 | 99.8% | 656.59 g/mol | C32H32O15 | 1 MG
SDP

Supplier:  Medchemexpress LLC HY1018951MG

Encompass_Preferred

Fluorescein di(β-D-galactopyranoside) is a fluorogenic substrate for β-galactosidase that releases fluorescein upon enzymatic cleavage, producing green fluorescence used to detect β-galactosidase activity in cells by fluorescence microscopy and flow cytometry.

  • Generates green fluorescence suitable for microscopy and flow cytometry.
  • High reported purity for consistent assay performance.
  • Sensitive detection of β-galactosidase activity and LacZ expression.
  • Soluble in common solvents such as DMSO for assay preparation.
  • Store at -20 °C and protect from light to preserve activity.

Catalog No. 50-002-49248


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.