Learn More
Abcam Fluorescein diacetate (FDA), 1G
Supplier:  Abcam AB2750871G
Encompass_Preferred
MW 416.4 Da, Purity >98%. Widely-used fluorogenic substrate for esterase activity. Upon cleavage fluorescein is released producing a green fluorescence that can be read using standard FITC filter sets. Common viability stain for plant cells, protoplasts and tissues. .
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-996-066