Learn More
Medchemexpress LLC Fluorescein hydrazide | 78044-20-9 | MFCD30470113 | C20H14N2O4 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY151644100MG
Encompass_Preferred
Fluorescein hydrazide is a sensitive fluorescent probe that induces noticeable color and fluorescence changes in the presence of Hg2+ and Co2+. It is prepared by the reaction of fluorescein and hydrazine. This probe exhibits an excitation wavelength of 508 nM and an emission wavelength of 531 nM.
  • Sensitive fluorescent probe for Hg2+ and Co2+.
  • Induces obvious color and fluorescence changes.
  • Excitation wavelength of 508 nM and emission wavelength of 531 nM.
  • Forms a complex with Hg2+, triggering structural changes.
  • Useful for detecting mercury pollution in water, food, and biological samples.
  • Applicable in environmental monitoring, food safety, and biomedicine.
Catalog No. 50-003-72053