Learn More
Medchemexpress LLC Fluorescein hydrazide | 78044-20-9 | MFCD30470113 | C20H14N2O4 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY15164450MG
Encompass_Preferred
Fluorescein hydrazide is a sensitive fluorescent probe designed to detect Hg2+ and Co2+. It exhibits clear color and fluorescence changes upon interaction with these ions. This compound is synthesized through the reaction of fluorescein and hydrazine, featuring an excitation wavelength of 508 nM and an emission wavelength of 531 nM.
  • Forms a complex with Hg2+, altering absorption and fluorescence spectra
  • Applicable for detecting mercury pollution in water, food, and biological samples
  • Useful in environmental monitoring, food safety, and biomedicine
Catalog No. 50-003-73216