Learn More
Sigma Aldrich Fine Chemicals Biosciences Fluorescein isothiocyanate5G
Supplier:  Sigma Aldrich Fine Chemicals Biosciences FD150S5G
Encompass
Dextran is a polymer of anhydroglucose. It is composed of approximately 95% alpha-D-(166) linkages. The remaining (163) linkages account for the branching of dextran. Conflicting data on the branch lengths implies that the average branch length is less than three glucose units. However other methods indicate branches of greater than 50 glucose units exist. Native dextran has been found to have a molecular weight (MW) in the range of 9 million to 500 million. Lower MW dextrans will exhibit slightly less branching and have a more narrow range of MW distribution. Dextrans with MW greater than 10 000 behave as if they are highly branched. As the MW increases dextran molecules attain greater symmetry. Dextrans with MW of 2 000 to 10 000 exhibit the properties of an expandable coil. At MW below 2 000 dextran is more rod-like. The MW of dextran is measured by one or more of the following methods low angle laser light scattering size exclusion chromatography copper-complexation and anthr
Catalog No. 50-290-7709