Learn More
Waters Corp Waters Fluorescence Detector Standard, Liquid, 10 mL, 24 Month Shelf Life
Supplier: Waters Corp 700003694
This Fluorescence Detector Standard is designed for system assessment of High-Performance Liquid Chromatography (HPLC) systems equipped with a fluorescence detector.
- Used for system assessment of HPLC with fluorescence detector.
- Composed of 5.0 pg/μL Anthracene ± 1% in 20/80 LC grade water / LC grade Acetonitrile.
- Includes a Certificate of Analysis.
- Formulated as a liquid.
- Recommended storage condition: room temperature.
- Shelf life: 24 months.
- Shipping condition: ambient.
- Volume: 10 mL.
- Type: reference standard.
- Application: system performance.
- This product contains hazardous material which requires special freight handling, and additional charges may apply.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.