Learn More
Selleck Chemical LLC Fluorescent Brightener 28 E4034-250MG
SDP
Supplier:  Selleck Chemical LLC E4034250MG
Encompass_Preferred
Fluorescent Brightener 28 (Tinopal UNPA-GX) is a viability stain used for identification of structure chitin and fungi It is also used as a fluorescent brightening agent for cellulose and polyamide fabrics paper and in detergents and soaps
Catalog No. 50-000-39080