Learn More
Medchemexpress LLC Fluorescent DOTAP (NBD-DOTAP) | 1010076-97-7 | 96.0% | 710.34 g·mol⁻¹ | C36H60ClN5O7 | 5 MG
Supplier: Medchemexpress LLC HY1437025MG
Fluorescent DOTAP (NBD-DOTAP) is an NBD-labeled cationic lipid used for nucleic acid and protein delivery, membrane labeling, and nanocarrier tracking. The NBD fluorophore provides visible-range excitation and emission suitable for imaging and formulation research. The compound is supplied as a solid and should be protected from light and stored under the temperature conditions recommended for solvent or bulk storage.
- NBD-labeled cationic lipid for delivery and imaging.
- Suitable for nucleic acid and protein transfection studies.
- Visible-range fluorescence (approx. 463/536 nm) for tracking.
- High purity for research applications.
- Stable solid form; protect from light and store under recommended conditions.