Learn More
Hach Company Fluoride ISA 1 L
Supplier:  Hach Company 2829053
Encompass
Fluoride Ionic Strength Adjustor (ISA)-1 LFluoride Total Ionic Strength Adjustor Buffer Solution (TISAB) for fluoride determinations by electrode methods Eliminates interferences from aluminum and iron
Catalog No. 50-003-04260