Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fluorochrome Inc Fluoro-Gold, retrograde axonal tracer, 20 mg, golden-yellow crystalline powder, ≥99% purity, CAS 223769-64-0

Supplier:  Fluorochrome Inc FLUOROGOLD20MG

Encompass

Fluoro-Gold is a fluorescent retrograde axonal tracer and histochemical stain, renowned for its effectiveness in neuroanatomical studies. This product stands out due to its intense fluorescence and extensive dendrite filling, making it ideal for long-term labeling and comprehensive neuronal pathway analysis. Fluoro-Gold ensures significant retrograde axonal transport and exhibits high stability across various histochemical techniques.

  • CAS Number: 223769-64-0
  • Molecular Weight: 532.6 daltons
  • Molecular Formula: C16H16N4O•2CH4O3S
  • Appearance: Golden-yellow crystalline powder
  • Purity: ≥99%
  • Primary Application: Retrograde axonal tracer
  • Secondary Application: Histochemical stain
  • Storage (Dry): Light-tight container at 4 °C
  • Shelf Life (Dry): More than 1 year
  • Solubility: Distilled water, 0.9% saline
  • Recommended Survival Time: 4-14 days

Catalog No. NC1363013


May include imposed supplier surcharges.
Only null left
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.