Learn More
Medchemexpress LLC Fluorocholine chloride | 459424-38-5 | MFCD07995053 | >95.0% | 157.61 | C5H13ClFNO | 250 MG

Supplier: Medchemexpress LLC HY107351250MG
Fluorocholine chloride is a quaternary ammonium biochemical reagent used in life-science research and biochemical assays. It is supplied in solid and solution forms and is intended for research use. The compound is water- and DMSO-soluble and is typically stored protected from moisture at refrigerated or frozen conditions depending on formulation.
- Quaternary ammonium reagent for biochemical assays and research use.
- Soluble in DMSO and water at approximately 100 mg/mL (may require ultrasonic).
- Storage: 4°C sealed; in solvent -80°C (6 months) or -20°C (1 month) when sealed and dry.
- Molecular weight 157.61 g/mol.
- CAS number 459424-38-5.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.