Learn More
Medchemexpress LLC Fluorocholine (chloride) | 459424-38-5 | MFCD07995053 | ≥98.0% | 157.61 | C5H13ClFNO | 1 G
SDP
Supplier:  Medchemexpress LLC HY1073511G
Encompass_Preferred
Fluorocholine (chloride) is a small-molecule biochemical reagent supplied as the chloride salt of fluorocholine, intended for life-science research such as tracer preparation and biochemical assays. It is a hydrophilic quaternary ammonium compound with demonstrated solubility in water and DMSO and defined storage recommendations for stability.
  • Used as a reagent or tracer in biochemical assays and imaging studies.
  • Molecular formula C5H13ClFNO, molecular weight 157.61 g/mol.
  • High solubility in water and DMSO (≈100 mg/mL); may require sonication.
  • Store sealed, away from moisture; in solvent: -80°C (6 months) or -20°C (1 month).
  • Available in lab-scale quantities suitable for research use.
  • Not intended for human or clinical use; for research use only.
Catalog No. 50-002-41489