Learn More
Starna Cells Inc FLUOROMETER MICROCELL SOG 3MM
SDP
Supplier:  Starna Cells Inc 33.45SOG3
Encompass_Preferred
Micro Square Special Optical Glass Fluorometer Cell, 45mm Height, 3mm Light Path
Catalog No. NC9040289