Learn More
Cytiva FluoroTrans W PVDF Transfer Membrane - 10 x 15 cm sheets (10/pkg)
Supplier:  Cytiva BSP0157
Encompass
Sensitive Protein Detection with Low Background and Low Burnthrough-Naturally hydrophobic polyvinylidene fluoride is ideal for a variety of protein-analysis applications. FluoroTrans media are white, microporous solid phase supports that bind proteins tenaciously via hydrophobic interactions. -High sensitivity for small peptides.-Typically absorbs 50% more protein than nylon or nitrocellulose.-FluoroTrans PVDF membrane is optimized for N-terminal protein sequencing. -FluoroTrans W membrane is optimized for Western blot applications. This membrane allows for sensitive protein detection with low background and low protein burnthrough. Immobilized proteins can be visually detected with Coomassie blue, Amido Black, Ponceau S, and colloidal gold. This product is not approved for use in medical, clinical, surgical or other patient applications.
Catalog No. 50-206-3439