Learn More
Abcam 5-Fluorouridine 5'-O-ß-D-galactopyranoside, 5MG
Supplier:  Abcam AB2738925MG
Encompass_Preferred
MW 424.33 Da, Purity =98%. Potent anti-metabolite. Acts via thymidylate synthetase inhibition or incorporation into RNA *in vivo*. Analog of 5-fluorouridine. Has galactose conjugated to the active site. Enzymatic or chemical hydrolysis of the galactoside group releases the active anti-metabolite. Useful for cell selection through lacZ specific release of 5-fluorouridine in recombinant cells or tissues.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-996-077