Learn More
Apexbio Technology LLC Fluoxetine 54910-89-3 100mg
SDP
Supplier:  Apexbio Technology LLC BA3590100
Encompass_Preferred
Fluoxetine (CAS 54910-89-3) is a selective serotonin reuptake inhibitor (SSRI) that acts by inhibiting the reabsorption of serotonin (5-hydroxytryptamine 5-HT) into presynaptic neurons thereby increasing synaptic serotonin levels This modulation of serotonergic neurotransmission makes fluoxetine a valuable tool in neuropharmacological studies focused on mood regulation psychiatric disorders and central nervous system function It is widely used in preclinical research to investigate the molecular and cellular mechanisms underlying depression anxiety and other mood-related conditions
Catalog No. 50-000-87830