Learn More
TARGETMOL CHEMICALS INC FLUOXETINE HYDROCHLORIDE 200MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T0450L200MG
Encompass_Preferred
Also available in 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Fluoxetine hydrochloride (Lilly110140) is the first highly specific serotonin uptake inhibitor. It is used as an antidepressant and often has a more acceptable side-effects profile than traditional antidepressants. purity: 99%
Catalog No. 50-258-1785