Learn More
Medchemexpress LLC Fluoxetine hydrochloride | 56296-78-7 | 100.0% | C17H19ClF3NO | 100 MG
Supplier: Medchemexpress LLC HYB0102A100MG
Fluoxetine hydrochloride (LY 110140) is an antidepressant and a selective serotonin reuptake inhibitor. It exhibits potent inhibitory activity against human SERT (serotonin reuptake transporter) in HEK293 cells with an IC50 of 150 nM. This product is for research use only.
- Selective serotonin reuptake inhibitor
- Potent inhibitory activity against human SERT in HEK293 cells (IC50: 150 nM)
- Blocks downregulation of cell proliferation in hippocampal cells
- Increases newborn cells in the dentate gyrus of adult rat hippocampus
- Enhances neurogenesis-dependent long-term potentiation (LTP)
- Increases extracellular norepinephrine and dopamine levels in the prefrontal cortex
- Reverses escape latency deficits in rat models of inescapable shock