Learn More
Medchemexpress LLC Fluoxetine hydrochloride | 56296-78-7 | 100.0% | C17H19ClF3NO | 500 MG
SDP
Supplier:  Medchemexpress LLC HYB0102A500MG
Encompass_Preferred
Fluoxetine hydrochloride is an antidepressant and a selective serotonin reuptake inhibitor, intended for research use only.
  • Acts as an antidepressant and selective serotonin reuptake inhibitor.
  • Blocks downregulation of cell proliferation in hippocampal cells.
  • Increases neurogenesis in the hippocampus and prelimbic cortex.
  • Enhances neurogenesis-dependent long-term potentiation.
  • Increases norepinephrine and dopamine extracellular levels.
  • Reverses deficits in escape latency in animal models.
Catalog No. 50-004-32308