Learn More
Apexbio Technology LLC Flupenthixol dihydrochloride 2413-38-9 10mg
SDP
Supplier:  Apexbio Technology LLC B757810
Encompass_Preferred
Flupenthixol dihydrochloride (CAS 2413-38-9) is a thioxanthene-derived small molecule primarily developed as an antipsychotic agent It functions as a dopamine D2 receptor antagonist modulating dopaminergic neurotransmission in the central nervous system In vitro assays demonstrate minimum inhibitory concentrations (MICs) of 10 100 g/mL against most bacterial strains with inhibitory effects observed at lower concentrations for select species including Staphylococcus aureus and Vibrio cholerae In vivo studies show that flupenthixol significantly reduces bacterial load and increases survival in murine infection models Clinically it has been evaluated for the management of schizophrenia and mood disorders supporting its utility in neuropharmacological and antimicrobial research
Catalog No. 50-000-91083