Learn More
Medchemexpress LLC Fluphenazine decanoate | 5002-47-1 | 98.8% | 100 MG
SDP
Supplier:  Medchemexpress LLC HYB1904100MG
Encompass_Preferred
Fluphenazine decanoate is a CNS-penetrant dopamine D2 receptor inhibitor and a long-acting phenothiazine neuroleptic. It can be used for schizophrenia research.
  • CNS-penetrant dopamine D2 receptor inhibitor
  • Long-acting phenothiazine neuroleptic
  • Used for schizophrenia research
  • Shows activity against *T. gondii* in human fibroblast cell cultures with an IC50 value of 1.7 mM
Catalog No. 50-004-10475