Learn More
Medchemexpress LLC Fluphenazine decanoate | 5002-47-1 | 98.8% | 50 MG
SDP
Supplier:  Medchemexpress LLC HYB190450MG
Encompass_Preferred
Fluphenazine decanoate is a CNS-penetrant dopamine D2 receptor inhibitor and a long-acting phenothiazine neuroleptic. It can be used for schizophrenia research.
  • CNS-penetrant dopamine D2 receptor inhibitor.
  • Long-acting phenothiazine neuroleptic.
  • Can be used for schizophrenia research.
  • Shows activity against T. gondii in human fibroblast cell cultures with an IC50 value of 1.7 mM.
  • Increases sensitivity to dopamine in animal models.
  • Induces mouth movements (pharmacological model of human tardive dyskinesia) in animal models.
  • Shows antifertility effects by inducing hyperprolactinemia in animal models.
Catalog No. 50-004-09654