Learn More
Medchemexpress LLC Fluphenazine-N-2-chloroethane hydrochloride | 3892-78-2 | 95.5% | 528.89 g/mol | C22H27Cl3F3N3S | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1337825MG
Encompass_Preferred
Fluphenazine-N-2-chloroethane hydrochloride is a phenothiazine-derived irreversible calmodulin antagonist used in biochemical and pharmacological research. Supplied as the hydrochloride salt, it is intended for in vitro studies of calmodulin modulation and related signaling pathways.
  • Irreversible calmodulin antagonist for biochemical assays.
  • Hydrochloride salt form for improved solubility.
  • White to off-white solid appearance for convenient handling.
  • High purity (95.5%) for reliable experimental results.
  • Stable when stored sealed, away from moisture and light.
Catalog No. 50-002-62700