Learn More
Cayman Chemical FluphenazInhydrochlorIde 500mg
SDP
Supplier:  Cayman Chemical 23555500 MG
Encompass_Preferred
A D1 and D2 receptor antagonist (Kis = 5 and 0.63 nM, respectively); binds to histamine H1, serotonin 5-HT2A, and 5-HT2B receptors (Kis = 45, 17, and 82 nM, respectively); acts as an inverse agonist at H1 and 5-HT2A receptors; a typical antipsychotic compound that induces extrapyramidal symptoms in monkeys
Catalog No. 50-296-4502