Learn More
Abcam Flupirtine maleate, Neuronal potassium channel opener, 25MG
Supplier:  Abcam AB14243225MG
Encompass_Preferred
MW 420.4 Da, Purity >99%. Neuronal potassium channel opener. Activates Kv7.2 channels and potently activates the inwardly rectifying potassium current (KIR; EC₅₀ = 600 nM). Centrally acting, non-opiate analgesic. Neuroprotective and antiapoptotic agent. Also has muscle relaxant properties. Active in vivo.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6237