Learn More
Medchemexpress LLC Fluralaner impurity 2 | 864725-62-2 | MFCD34574502 | C18H12Cl2F3NO3 | 500 MG

Supplier: Medchemexpress LLC HY45298500MG
Fluralaner impurity 2 is a chemical compound characterized as an impurity of Fluralaner. It has a molecular weight of 418.19 and a chemical formula of C18H12Cl2F3NO3. This compound appears as a solid and is intended for research purposes only.
- Impurity of Fluralaner
- Molecular weight of 418.19
- Chemical formula C18H12Cl2F3NO3
- Appears as a solid
- For research purposes only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.