Learn More
Cayman Chemical -FlurbIprofen 10g
SDP
Supplier:  Cayman Chemical 7025010 G
Encompass_Preferred
A non-selective COX inhibitor (IC50s = 0.04 and 0.51 μM for COX-1 and COX-2, respectively); reduces carrageenan-induced hind paw edema and yeast-induced fever in rats from 0.3-4.8 mg/kg; reduces plasma fibrinogen levels and arthritic score in a rat model of adjuvant-induced arthritis; reduces tumor weight and prostaglandin production and increases survival in a WHT-NC mouse xenograft model at 5 mg/kg
Catalog No. 50-289-6874