Learn More
Medchemexpress LLC Flurbiprofen | 5104-49-4 | 100.0% | 244.26 g/mol | C15H13FO2 | 1 ML

Supplier: Medchemexpress LLC HY1058210MM 1ML
Flurbiprofen is a nonsteroidal anti-inflammatory research reagent that functions as a non-selective cyclooxygenase (COX) inhibitor. It is used in studies of inflammation, pain signaling, and certain cancer research applications. Supplied as a 10 mM solution (1 mL), the material is intended for laboratory research.
- Acts as a non-selective COX inhibitor.
- Used for inflammation, analgesia, and cancer-related research.
- High purity (99.98% by LCMS).
- Molecular weight 244.26 g/mol; formula C15H13FO2.
- CAS 5104-49-4.
- Store in solvent at -80°C (long term) or -20°C (up to 1 year).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.