Learn More
Apexbio Technology LLC Flurbiprofen 5104-49-4 100mg
SDP
Supplier:  Apexbio Technology LLC A8434100
Encompass_Preferred
Flurbiprofen (CAS 5104-49-4) is a nonsteroidal anti-inflammatory compound that functions as an inhibitor of cyclooxygenase (COX) enzymes thereby reducing the synthesis of prostaglandins involved in inflammatory processes This small molecule is frequently employed in biomedical research to study the molecular mechanisms underlying inflammation and to assess potential therapeutic strategies for inflammatory conditions such as osteoarthritis and rheumatoid arthritis Its well-characterized pharmacological profile makes it a valuable tool for preclinical studies investigating pathways associated with pain and inflammation
Catalog No. 50-000-96251