Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Flurbiprofen | 5104-49-4 | MFCD00079303 | 100.0% | 244.26 g/mol | C15H13FO2 | 1 G
SDP

Supplier:  Medchemexpress LLC HY105821G

Encompass_Preferred

Flurbiprofen is a nonsteroidal anti-inflammatory drug (NSAID) supplied as a research reagent and cyclooxygenase (COX) inhibitor. It is used in biochemical and pharmacological studies of inflammation, pain, and prostaglandin-mediated signaling.

  • High purity (99.98%).
  • Solid, white to light yellow appearance.
  • Molecular weight 244.26 g/mol.
  • Chemical formula C15H13FO2.
  • Contains CAS number 5104-49-4 for substance identification.
  • Available in 1 g and larger solid pack sizes for laboratory use.
  • Suitable for in vitro COX inhibition and pharmacology assays.

Catalog No. 50-002-43138


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.