Learn More
Selleck Chemical LLC Flurbiprofen Axetil
SDP
Supplier:  Selleck Chemical LLC S64575MG
Encompass_Preferred
Flurbiprofen Axetil is one of the commonly used nonsteroidal anti-inflammatory drugs (NSAIDs) that nonselectively inhibits cyclooxygenase
Catalog No. 50-000-04787