Learn More
Medchemexpress LLC Flurbiprofen axetil | 91503-79-6 | MFCD00900049 | 99.3% | 330.35 g/mol | C19H19FO4 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1014815MG
Encompass_Preferred
Flurbiprofen axetil is a nonsteroidal anti-inflammatory prodrug of flurbiprofen that acts as a non-selective cyclooxygenase (COX) inhibitor with analgesic, anti-inflammatory, and reported neuroprotective effects. Supplied as a colorless to off-white oil for research use only; typical purity approximately 99.3%.
  • Non-selective COX inhibitor with analgesic and anti-inflammatory activity.
  • Demonstrates neuroprotective effects in preclinical models.
  • High purity (~99.3%) suitable for research applications.
  • Supplied as a colorless to off-white oil for easy formulation.
  • Available in multiple small pack sizes for preclinical studies.
Catalog No. 50-002-47961