Learn More
Cayman Chemical FlurbIprofen-d3 500ug
SDP
Supplier:  Cayman Chemical 25277500 UG
Encompass_Preferred
An internal standard for the quantification of flurbiprofen by GC- or LC-MS
Catalog No. 50-293-2640