Learn More
Medchemexpress LLC Flurbiprofen-d5 | 215175-76-1 | MFCD27992097 | 99.8% | 249.29 g/mol | C15H8D5FO2 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY10582S15MG
Encompass_Preferred
Flurbiprofen-d5 is the deuterium-labeled isotopologue of flurbiprofen used as a stable isotope reference standard for analytical and mass spectrometry applications. It functions as an internal standard for quantitative LC-MS/MS assays and is provided in small, accurately weighed packages for method development and validation.
  • Deuterated internal standard for LC-MS quantitation.
  • High chemical purity: 99.8%.
  • Molecular formula C15H8D5FO2 and molecular weight 249.29 g/mol.
  • Supplied in small, calibrated quantities for accurate dosing.
  • Suitable for method development, validation, and quantitative analysis.
Catalog No. 50-002-41863