Learn More
Alkali Scientific Flurprimidol, 25 Mg
SDP
Supplier:  Alkali Scientific URP001CP
Encompass_Preferred
Flurprimidol is a plant growth regulator used to control the growth of plants by inhibiting gibberellin biosynthesis. Available in 25 mg amounts, it is used in laboratory settings for controlling excessive plant growth and promoting compact plant forms. Flurprimidol is often used in plant tissue culture experiments to regulate the growth of plant tissues and enhance the success of culture protocols. By inhibiting gibberellin, flurprimidol helps maintain desirable plant morphology, making it useful for both research and commercial plant breeding. It is widely used in controlled environments to achieve optimal plant size and shape, particularly in ornamental and agricultural crops.
Catalog No. 50-006-60568