Learn More
FUJIFILM BIOSCIENCES INC Fluthiacet-methyl Standard
Supplier:  FUJIFILM BIOSCIENCES INC 06204461
Encompass_Preferred
High-purity standard of Fluthiacet-methyl 99 0% GC 98 0% qNMR in white crystalline powder form intended for use in pesticide residue analysis Stable when stored at 2 10 C protected from light
Catalog No. 50-004-50358