Learn More
Medchemexpress LLC Fluticasone | 90566-53-3 | 98.7% | 5 MG
SDP
Supplier:  Medchemexpress LLC HYB06035MG
Encompass_Preferred
Fluticasone is an inhaled corticosteroid used for respiratory research. It functions as a Smo agonist with an IC50 value of 99 nM, and also activates Hedgehog signaling. It stimulates the proliferation of primary neuronal stem or precursor cells. This product is for research use only and not intended for patient use.
  • Acts as a Smo agonist with an IC50 value of 99 nM.
  • Activates Hedgehog signaling.
  • Stimulates proliferation of primary neuronal stem or precursor cells.
  • Inhibits U2OS cell growth.
  • Reduces HRV-induced mucin production.
  • Suppresses rhinovirus-induced airway inflammation.
Catalog No. 50-004-08359