Learn More
Selleck Chemical LLC fluticasone furoate
SDP
Supplier:  Selleck Chemical LLC S64875MG
Encompass_Preferred
Fluticasone furoate (GW685698) is an inhaled corticosteroid/long-acting beta-agonist combination with a prolonged bronchodilator duration of action It is used for the treatment of non-allergic and allergic rhinitis administered by a nasal spray
Catalog No. 50-000-03453