Learn More
Medchemexpress LLC Fluticasone furoate | 397864-44-7 | C27H29F3O6S | 50 MG
SDP
Supplier:  Medchemexpress LLC HY1523450MG
Encompass_Preferred
Fluticasone furoate is a topical, intranasal, synthetic trifluorinated corticosteroid with potent anti-inflammatory and anti-asthmatic activity. It exhibits low systemic exposure and is primarily metabolized in the liver. It holds potential for the treatment of allergic rhinitis.
  • Enhanced-affinity synthetic trifluorinated corticosteroid
  • Potent anti-inflammatory and anti-asthmatic activity
  • Low systemic exposure
  • Potential for allergic rhinitis treatment
  • Provided as a nasal spray for topical administration
  • Inhibits tumor necrosis factor synthesis and action
  • Effective in preventing damage to cultured human lung epithelial cells
  • High plasma protein binding (99.4%)
  • Extensive first-pass metabolism by CYP3A4
  • Mainly excreted in the feces
Catalog No. 50-003-72587